Assay Details
Target Gene Details
Entrez Gene ID: | 5187 |
Gene Name: | period circadian clock 1 |
Gene Aliases: |
PER, RIGUI, hPER |
Location: |
Chr.17:8140470-8156405 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PER1 | NM_002616.2 | 23 | 4167 | NP_002607.2 |
| XM_005256689.1 | 23 | 4167 | XP_005256746.1 | |
| XM_005256690.1 | 22 | 3987 | XP_005256747.1 | |
| AB002107.1 | BAA22633.1 | |||
| AB088477.1 | 23 | 4109 | ||
| AF022991.1 | 23 | 4116 | AAC51765.1 | |
| AK291061.1 | 23 | 4127 | ||
| BC137346.1 | 22 | 4076 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv518983 | Chr.17:8124275 - 8147661 on Build GRCh38 | Loss |
|
| nsv527475 | Chr.17:8140435 - 8189376 on Build GRCh38 | Loss |
|
| nsv1964 | Chr.17:8140093 - 8175220 on Build GRCh38 | Insertion |
|
| nsv833352 | Chr.17:8029719 - 8187575 on Build GRCh38 | Loss |
|
| nsv457661 | Chr.17:8106586 - 8147661 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map