Assay Details
Target Gene Details
Entrez Gene ID: | 26279 |
Gene Name: | phospholipase A2 group IID |
Gene Aliases: |
PLA2IID, SPLASH, sPLA2-IID, sPLA2S |
Location: |
Chr.1:20112650-20119566 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLA2G2D | NM_001271814.1 | NP_001258743.1 | ||
| NM_012400.3 | NP_036532.1 | |||
| AF112982.1 | AAD51390.1 | |||
| AF188625.1 | AAF09020.1 | |||
| AK290406.1 | ||||
| AK301396.1 | ||||
| AK310156.1 | ||||
| AK311903.1 | ||||
| BC025706.1 | AAH25706.1 | |||
| DC295429.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv1003821 | Chr.1:20104576 - 20116485 on Build GRCh38 | Deletion |
|
| nsv545731 | Chr.1:19989106 - 20776306 on Build GRCh38 | Gain |
|
| nsv1003802 | Chr.1:20057935 - 20154347 on Build GRCh38 | Loss |
|
| nsv5520 | Chr.1:20071053 - 20115978 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map