Assay Details
Target Gene Details
Entrez Gene ID: | 64969 |
Gene Name: | mitochondrial ribosomal protein S5 |
Gene Aliases: |
MRP-S5, S5mt |
Location: |
Chr.2:95085391-95122027 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPS5 | NM_001321995.1 | NP_001308924.1 | ||
| NM_001321996.1 | NP_001308925.1 | |||
| NM_001321997.1 | NP_001308926.1 | |||
| NM_031902.4 | NP_114108.1 | |||
| NR_136152.1 | ||||
| NR_136153.1 | ||||
| AB049940.1 | BAB40993.1 | |||
| AK058160.1 | BAB71695.1 | |||
| AK295812.1 | ||||
| BC000219.2 | AAH00219.2 | |||
| BC014172.2 | AAH14172.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1160825 | Chr.2:95050220 - 95445574 on Build GRCh38 | Duplication |
|
| dgv4007n100 | Chr.2:94662150 - 95248719 on Build GRCh38 | Gain |
|
| esv3591662 | Chr.2:94986856 - 95146757 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map