Assay Details
Target Gene Details
Entrez Gene ID: | 2532 |
Gene Name: | atypical chemokine receptor 1 (Duffy blood group) |
Gene Aliases: |
CCBP1, CD234, DARC, Dfy, FY, GPD, GpFy, WBCQ1 |
Location: |
Chr.1:159204013-159206500 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACKR1 | NM_002036.3 | 1 | 721 | NP_002027.2 |
| AK291593.1 | 1 | 721 | ||
| BX537430.1 | 2 | 618 | CAD97672.1 | |
| CD701182.1 |
More Information
Set Membership: |
|

Genomic Map