Assay Details
Target Gene Details
Entrez Gene ID: | 1267 |
Gene Name: | 2',3'-cyclic nucleotide 3' phosphodiesterase |
Gene Aliases: |
CNP1 |
Location: |
Chr.17:41966741-41977736 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CNP | NM_033133.4 | 2 | 316 | NP_149124.3 |
| XM_006721701.3 | 2 | 340 | XP_006721764.1 | |
| XM_011524340.2 | 2 | 301 | XP_011522642.1 | |
| AK026876.1 | 2 | 273 | ||
| AK098384.1 | 2 | 262 | ||
| AK294179.1 | ||||
| AK295351.1 | 2 | 266 | ||
| AK308696.1 | ||||
| BC001362.2 | 2 | 253 | AAH01362.1 | |
| BC006392.2 | 2 | 275 | AAH06392.1 | |
| BC011046.1 | 2 | 237 | AAH11046.1 | |
| BC028040.1 | 2 | 277 | AAH28040.1 | |
| CB988051.1 | 2 | 343 | ||
| M19650.1 | 1 | 125 | AAA35704.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509658 | Chr.17:41927491 - 42002888 on Build GRCh38 | Insertion |
|
| nsv953897 | Chr.17:41966883 - 41977382 on Build GRCh38 | Deletion |
|
| nsv2056 | Chr.17:41923301 - 41975161 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map