Assay Details
Target Gene Details
Entrez Gene ID: | 6370 |
Gene Name: | C-C motif chemokine ligand 25 |
Gene Aliases: |
Ckb15, SCYA25, TECK |
Location: |
Chr.19:8052318-8062663 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCL25 | NM_001201359.1 | 6 | 796 | NP_001188288.1 |
| NM_005624.3 | 6 | 799 | NP_005615.2 | |
| XM_011528177.2 | 7 | 1000 | XP_011526479.1 | |
| XM_017027119.1 | 7 | 966 | XP_016882608.1 | |
| XM_017027120.1 | 7 | 895 | XP_016882609.1 | |
| XM_017027121.1 | 7 | 841 | XP_016882610.1 | |
| XM_017027122.1 | 7 | 975 | XP_016882611.1 | |
| AI313430.1 | 1 | 206 | ||
| AI961745.1 | 1 | 211 | ||
| AK292662.1 | ||||
| U86358.1 | 5 | 688 | AAB69981.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833734 | Chr.19:7960566 - 8138311 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map