Assay Details
Target Gene Details
Entrez Gene ID: | 40 |
Gene Name: | acid sensing ion channel subunit 2 |
Gene Aliases: |
ACCN, ACCN1, ASIC2a, BNC1, BNaC1, MDEG, hBNaC1 |
Location: |
Chr.17:33013087-34156806 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASIC2 | NM_001094.4 | NP_001085.2 | ||
| NM_183377.1 | NP_899233.1 | |||
| AK127748.1 | ||||
| AL834182.1 | CAD38879.1 | |||
| U57352.1 | AAB49182.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv510705 | Chr.17:32841518 - 33023081 on Build GRCh38 | Deletion |
|
| nsv833417 | Chr.17:32843798 - 33018580 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map