Assay Details
Target Gene Details
Entrez Gene ID: | 8974 |
Gene Name: | prolyl 4-hydroxylase subunit alpha 2 |
Gene Aliases: |
- |
Location: |
Chr.5:132190147-132227863 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| P4HA2 | NM_001017973.1 | 7 | 1420 | NP_001017973.1 |
| NM_001017974.1 | 7 | 948 | NP_001017974.1 | |
| NM_001142598.1 | 8 | 1084 | NP_001136070.1 | |
| NM_001142599.1 | 8 | 1084 | NP_001136071.1 | |
| NM_004199.2 | 7 | 1420 | NP_004190.1 | |
| XM_005272116.4 | 7 | 948 | XP_005272173.1 | |
| XM_005272117.4 | 8 | 1106 | XP_005272174.1 | |
| XM_005272118.4 | 8 | 1235 | XP_005272175.1 | |
| XM_005272119.4 | 8 | 1425 | XP_005272176.1 | |
| XM_005272120.4 | 8 | 1205 | XP_005272177.1 | |
| XM_006714728.3 | 8 | 1482 | XP_006714791.1 | |
| XM_006714729.3 | 8 | 1106 | XP_006714792.1 | |
| XM_006714730.3 | 8 | 1482 | XP_006714793.1 | |
| XM_011543705.2 | 6 | 874 | XP_011542007.1 | |
| XM_017010011.1 | 8 | 1205 | XP_016865500.1 | |
| XM_017010012.1 | 7 | 948 | XP_016865501.1 | |
| AY358970.1 | 7 | 1022 | AAQ89329.1 | |
| BC017062.1 | 7 | 1049 | ||
| BC035813.1 | 7 | 917 | AAH35813.1 | |
| BX647924.1 | 7 | 940 | CAI46066.1 | |
| U90441.1 | 7 | 1043 | AAB71339.1 |
More Information
Set Membership: |
|

Genomic Map