Assay Details
Target Gene Details
Entrez Gene ID: | 166979 |
Gene Name: | cell division cycle 20B |
Gene Aliases: |
G6VTS76519 |
Location: |
Chr.5:55112971-55173177 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDC20B | NM_001145734.2 | NP_001139206.2 | ||
| NM_001170402.1 | NP_001163873.1 | |||
| NM_152623.2 | NP_689836.2 | |||
| XM_011543218.2 | XP_011541520.1 | |||
| AB077043.1 | BAC67701.1 | |||
| AB077044.1 | BAB97393.1 | |||
| AB081750.1 | BAC67702.1 | |||
| AB086378.1 | BAC67705.1 | |||
| AK095246.1 | BAC04508.1 | |||
| BC037547.1 | AAH37547.1 | |||
| BC136572.1 | ||||
| DA434860.1 | ||||
| DB237622.1 |
Target Gene Details
Entrez Gene ID: | 493869 |
Gene Name: | glutathione peroxidase 8 (putative) |
Gene Aliases: |
EPLA847, GPx-8, GSHPx-8, UNQ847 |
Location: |
Chr.5:55160118-55167301 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPX8 | NM_001008397.3 | 3 | 935 | NP_001008398.2 |
| NM_001306197.1 | 3 | 782 | NP_001293126.1 | |
| NM_001306198.1 | 2 | 673 | NP_001293127.1 | |
| NM_001306201.1 | 3 | 656 | NP_001293130.1 | |
| NR_131336.1 | 4 | 849 | ||
| NR_131337.1 | 4 | 791 | ||
| NR_131338.1 | 3 | 587 | ||
| NR_131340.1 | 3 | 925 | ||
| XM_006714631.2 | 3 | 901 | XP_006714694.1 | |
| AK074216.1 | 3 | 938 | BAB85019.1 | |
| AK298545.1 | 3 | 730 | ||
| AK307365.1 | 2 | 611 | ||
| AY358715.1 | 3 | 861 | AAQ89077.1 | |
| BC029424.1 | 3 | 894 | AAH29424.1 | |
| BG620710.1 | ||||
| BX537894.1 | 4 | 799 | ||
| BX647770.1 | 2 | 623 | ||
| CN390753.1 | ||||
| DA660205.1 | ||||
| HY174481.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830302 | Chr.5:55107550 - 55259351 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map