Assay Details
Target Gene Details
Entrez Gene ID: | 2863 |
Gene Name: | G protein-coupled receptor 39 |
Gene Aliases: |
- |
Location: |
Chr.2:132415972-132646596 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR39 | NM_001508.2 | 2 | 1683 | NP_001499.1 |
| AF034633.1 | 2 | 1214 | AAC26082.1 | |
| AK122643.1 | 2 | 1683 | ||
| AK123759.1 | 3 | 917 | ||
| AK314895.1 | 2 | 1440 | ||
| BC125046.1 | 2 | 1214 | ||
| BC125047.1 | 2 | 1203 | ||
| FJ348258.1 | 2 | 1214 |
Target Gene Details
Entrez Gene ID: | 116372 |
Gene Name: | LY6/PLAUR domain containing 1 |
Gene Aliases: |
LYPDC1, PHTS |
Location: |
Chr.2:132643280-132671579 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LYPD1 | NM_001077427.3 | 3 | 1362 | NP_001070895.1 |
| NM_001321234.1 | 4 | 1324 | NP_001308163.1 | |
| NM_001321235.1 | 3 | 1485 | NP_001308164.1 | |
| NM_144586.6 | 3 | 1451 | NP_653187.3 | |
| XM_011510546.2 | 4 | 2157 | XP_011508848.1 | |
| AK075487.1 | 3 | 1286 | BAC11647.1 | |
| AK094501.1 | 4 | 1227 | BAC04368.1 | |
| AK123029.1 | 4 | 1184 | BAC85518.1 | |
| AK130005.1 | 4 | 1163 | BAC85273.1 | |
| AY358628.1 | 3 | 1133 | AAQ88991.1 | |
| BC040046.1 | 1 | 67 | AAH40046.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv583155 | Chr.2:132495813 - 132657597 on Build GRCh38 | Loss |
|
| nsv583158 | Chr.2:132558519 - 133104527 on Build GRCh38 | Gain |
|
| nsv583152 | Chr.2:132369502 - 133631550 on Build GRCh38 | Gain |
|
| nsv1011421 | Chr.2:132137668 - 132924612 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map