Assay Details
Target Gene Details
Entrez Gene ID: | 3852 |
Gene Name: | keratin 5 |
Gene Aliases: |
CK5, DDD, DDD1, EBS2, K5, KRT5A |
Location: |
Chr.12:52514575-52520459 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRT5 | NM_000424.3 | 9 | 2152 | NP_000415.2 |
| AK093720.1 | 10 | 2018 | ||
| AK303972.1 | 10 | 1982 | ||
| BC024292.1 | 9 | 2076 | AAH24292.1 | |
| BC042132.1 | 9 | 2076 | AAH42132.1 | |
| BC071906.1 | 9 | 2071 | AAH71906.1 | |
| M19723.1 | 9 | 1743 | AAA36145.1 | |
| M21389.1 | 9 | 2380 | AAA36143.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv709 | Chr.12:52500235 - 52564462 on Build GRCh38 | Insertion |
|
| nsv558933 | Chr.12:52514689 - 52548578 on Build GRCh38 | Loss |
|
| nsv832412 | Chr.12:52390206 - 52545452 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map