Assay Details
Target Gene Details
Entrez Gene ID: | 4522 |
Gene Name: | methylenetetrahydrofolate dehydrogenase, cyclohydrolase and formyltetrahydrofolate synthetase 1 |
Gene Aliases: |
MTHFC, MTHFD |
Location: |
Chr.14:64388041-64460007 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 27 - Intron 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTHFD1 | NM_005956.3 | NP_005947.3 | ||
| AK095514.1 | 3 | 881 | ||
| AK302747.1 | ||||
| AK312361.1 | ||||
| BC001014.2 | AAH01014.2 | |||
| BC009806.2 | AAH09806.1 | |||
| BC050420.1 | AAH50420.1 | |||
| CA446032.1 | ||||
| J04031.1 | AAA59574.1 |
Target Gene Details
Entrez Gene ID: | 7597 |
Gene Name: | zinc finger and BTB domain containing 25 |
Gene Aliases: |
C14orf51, KUP, ZNF46 |
Location: |
Chr.14:64447172-64505213 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZBTB25 | NM_001304508.1 | NP_001291437.1 | ||
| XM_006720250.3 | XP_006720313.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2748779 | Chr.14:63663324 - 66595809 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map