Assay Details
Target Gene Details
Entrez Gene ID: | 1408 |
Gene Name: | cryptochrome circadian clock 2 |
Gene Aliases: |
HCRY2, PHLL2 |
Location: |
Chr.11:45847118-45883248 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRY2 | NM_001127457.2 | 9 | 1414 | NP_001120929.1 |
| NM_021117.3 | 9 | 1550 | NP_066940.2 | |
| AB014558.1 | 9 | 1454 | BAA31633.1 | |
| AK291260.1 | 9 | 1481 | ||
| AK294904.1 | 9 | 1408 | ||
| AK295627.1 | 5 | 950 | ||
| AK302865.1 | 9 | 1550 | ||
| BC035161.1 | 11 | 1813 | AAH35161.1 | |
| BC041814.1 | 9 | 1478 | AAH41814.1 | |
| EU219622.1 | 9 | 1473 |
More Information
Set Membership: |
|

Genomic Map