Assay Details
Target Gene Details
Entrez Gene ID: | 84000 |
Gene Name: | transmembrane protease, serine 13 |
Gene Aliases: |
MSP, MSPL, MSPS, TMPRSS11 |
Location: |
Chr.11:117900641-117929453 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMPRSS13 | NM_001077263.2 | 13 | 2651 | NP_001070731.1 |
| NM_001206789.1 | 12 | 2546 | NP_001193718.1 | |
| AB048797.1 | 12 | 2502 | ||
| BC114928.1 | 13 | 2570 | AAI14929.1 |
More Information
Set Membership: |
|

Genomic Map