Assay Details
Target Gene Details
Entrez Gene ID: | 120400 |
Gene Name: | neurexophilin and PC-esterase domain family member 1 |
Gene Aliases: |
FAM55A |
Location: |
Chr.11:114518946-114560244 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NXPE1 | NM_152315.3 | 6 | 1836 | NP_689528.2 |
| XM_011542594.2 | 8 | 2353 | XP_011540896.1 | |
| XM_011542595.2 | 9 | 2465 | XP_011540897.1 | |
| XM_011542596.2 | 9 | 2706 | XP_011540898.1 | |
| XM_011542597.2 | 8 | 2594 | XP_011540899.1 | |
| XM_011542598.2 | 7 | 2294 | XP_011540900.1 | |
| XM_011542599.2 | 7 | 2535 | XP_011540901.1 | |
| AI347274.1 | 1 | 81 | ||
| BC029049.1 | 6 | 1799 | AAH29049.1 | |
| BX648601.1 | 4 | 2199 |
Target Gene Details
Entrez Gene ID: | 120406 |
Gene Name: | neurexophilin and PC-esterase domain family member 2 |
Gene Aliases: |
FAM55B |
Location: |
Chr.11:114464276-114813702 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NXPE2 | XM_017017207.1 | XP_016872696.1 | ||
| XM_017017209.1 | XP_016872698.1 | |||
| XM_017017210.1 | XP_016872699.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759865 | Chr.11:114501563 - 114648403 on Build GRCh38 | Gain |
|

Genomic Map