Assay Details
Target Gene Details
Entrez Gene ID: | 5081 |
Gene Name: | paired box 7 |
Gene Aliases: |
HUP1, PAX7B, RMS2 |
Location: |
Chr.1:18631006-18748866 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PAX7 | NM_001135254.1 | NP_001128726.1 | ||
| NM_002584.2 | NP_002575.1 | |||
| NM_013945.2 | NP_039236.1 | |||
| BC121165.2 | ||||
| BC121166.2 | ||||
| DQ322591.1 | ABC48797.1 | |||
| X96743.1 | CAA65520.1 | |||
| Z35141.1 | CAA84513.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1006958 | Chr.1:18489777 - 18746432 on Build GRCh38 | Gain |
|
| nsv1008797 | Chr.1:18629403 - 18677739 on Build GRCh38 | Gain |
|
| nsv4965 | Chr.1:18605318 - 18638892 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map