Assay Details
Target Gene Details
Entrez Gene ID: | 25977 |
Gene Name: | NECAP endocytosis associated 1 |
Gene Aliases: |
EIEE21 |
Location: |
Chr.12:8082211-8097777 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NECAP1 | NM_015509.3 | NP_056324.2 | ||
| NR_024260.1 | ||||
| AK074858.1 | BAC11250.1 | |||
| AK074880.1 | BAC11264.1 | |||
| AK074923.1 | BAC11296.1 | |||
| AK075013.1 | BAC11352.1 | |||
| AK094805.1 | ||||
| AK290537.1 | ||||
| AK303675.1 | ||||
| AK314424.1 | ||||
| AL050272.1 | CAB43373.2 | |||
| BC002888.2 | AAH02888.1 | |||
| BC067367.1 | AAH67367.1 | |||
| BC110876.1 | AAI10877.1 | |||
| DA496321.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428270 | Chr.12:7704223 - 8471391 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map