Assay Details
Target Gene Details
Entrez Gene ID: | 9533 |
Gene Name: | RNA polymerase I subunit C |
Gene Aliases: |
AC40, HLD11, RPA39, RPA40, RPA5, RPAC1, RPC40, TCS3 |
Location: |
Chr.6:43517024-43562404 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLR1C | NM_001318876.1 | NP_001305805.1 | ||
| XM_011515000.2 | XP_011513302.1 | |||
| AF047441.1 | AAC39892.1 |
Target Gene Details
Entrez Gene ID: | 57510 |
Gene Name: | exportin 5 |
Gene Aliases: |
exp5 |
Location: |
Chr.6:43522330-43576075 on Build GRCh38 |
Assay Gene Location: | Within Exon 32 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| XPO5 | NM_020750.2 | 32 | 5126 | NP_065801.1 |
| AB033117.2 | 32 | 5065 | ||
| AF298880.1 | 32 | 4978 | AAG17907.1 | |
| AK001195.1 | 11 | 2459 | BAA91547.1 | |
| AK023431.1 | 19 | 2015 | BAB14572.1 | |
| AK025864.1 | 1 | 1399 | ||
| AK127513.1 | 3 | 3399 | ||
| BF108994.1 | 1 | 253 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv603000 | Chr.6:43397622 - 43524840 on Build GRCh38 | Loss |
|
| nsv818414 | Chr.6:43507736 - 43528924 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map