Assay Details
Target Gene Details
Entrez Gene ID: | 6369 |
Gene Name: | C-C motif chemokine ligand 24 |
Gene Aliases: |
Ckb-6, MPIF-2, MPIF2, SCYA24 |
Location: |
Chr.7:75810825-75823356 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCL24 | NM_002991.2 | NP_002982.2 | ||
| XM_011516459.2 | XP_011514761.1 | |||
| XM_011516460.2 | XP_011514762.1 | |||
| AJ223461.1 | CAA11383.1 | |||
| AK312216.1 | ||||
| BC069072.1 | AAH69072.1 | |||
| BC069391.1 | AAH69391.1 | |||
| U85768.1 | AAB51135.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1134897 | Chr.7:75809018 - 75822618 on Build GRCh38 | Tandem Duplication |
|
| nsv528969 | Chr.7:75074053 - 76920728 on Build GRCh38 | Gain |
|
| nsv518028 | Chr.7:75539242 - 75825134 on Build GRCh38 | Gain |
|
| dgv120n16 | Chr.7:75440769 - 76641919 on Build GRCh38 | Deletion |
|
| esv2764051 | Chr.7:75663839 - 75875518 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map