Assay Details
Target Gene Details
Entrez Gene ID: | 150590 |
Gene Name: | chromosome 2 open reading frame 15 |
Gene Aliases: |
- |
Location: |
Chr.2:99141613-99151487 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C2orf15 | NM_001317992.1 | 5 | 1186 | NP_001304921.1 |
| NM_144706.3 | 4 | 1090 | NP_653307.1 | |
| AA653513.1 | ||||
| BC021264.2 | 4 | 981 | AAH21264.1 |
Target Gene Details
Entrez Gene ID: | 80705 |
Gene Name: | testis specific 10 |
Gene Aliases: |
CEP4L, CT79 |
Location: |
Chr.2:98997261-99154962 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSGA10 | NM_025244.2 | NP_079520.1 | ||
| XM_006712781.2 | XP_006712844.1 | |||
| XM_011511937.1 | XP_011510239.1 | |||
| XM_017005040.1 | XP_016860529.1 | |||
| XM_017005041.1 | XP_016860530.1 | |||
| XM_017005044.1 | XP_016860533.1 | |||
| XM_017005047.1 | XP_016860536.1 | |||
| BX647157.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3591732 | Chr.2:99123694 - 99182166 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map