Assay Details
Target Gene Details
Entrez Gene ID: | 5422 |
Gene Name: | DNA polymerase alpha 1, catalytic subunit |
Gene Aliases: |
NSX, POLA, p180 |
Location: |
Chr.X:24693833-24996986 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLA1 | NM_016937.3 | 10 | 999 | NP_058633.2 |
| XM_005274552.3 | 10 | 1131 | XP_005274609.1 | |
| XM_006724499.2 | 9 | 941 | XP_006724562.1 | |
| XM_011545540.2 | 10 | 1086 | XP_011543842.1 | |
| XM_011545541.2 | 10 | 1087 | XP_011543843.1 | |
| XM_017029594.1 | 10 | 1087 | XP_016885083.1 | |
| XM_017029595.1 | 10 | 1087 | XP_016885084.1 | |
| XM_017029596.1 | 10 | 1087 | XP_016885085.1 | |
| BX648513.1 | 10 | 1024 | ||
| X06745.1 | 10 | 1000 | CAA29920.1 |
More Information
Set Membership: |
|

Genomic Map