Assay Details
Target Gene Details
Entrez Gene ID: | 2015 |
Gene Name: | adhesion G protein-coupled receptor E1 |
Gene Aliases: |
EMR1, TM7LN3 |
Location: |
Chr.19:6887549-6940453 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADGRE1 | NM_001256252.1 | NP_001243181.1 | ||
| NM_001256253.1 | NP_001243182.1 | |||
| NM_001256254.1 | NP_001243183.1 | |||
| NM_001256255.1 | NP_001243184.1 | |||
| NM_001974.4 | NP_001965.3 | |||
| XM_011527794.1 | XP_011526096.1 | |||
| AK131562.1 | BAD18695.1 | |||
| AK290401.1 | ||||
| AK291518.1 | ||||
| AK297003.1 | ||||
| AK297011.1 | ||||
| AK313495.1 | ||||
| BC059395.1 | AAH59395.1 | |||
| DA465986.1 | ||||
| X81479.1 | CAA57232.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv14e196 | Chr.19:6710054 - 6986820 on Build GRCh38 | Duplication |
|
| dgv136e55 | Chr.19:6820940 - 7112259 on Build GRCh38 | Gain |
|
| nsv833729 | Chr.19:6832389 - 6977151 on Build GRCh38 | Loss |
|
| dgv3421n100 | Chr.19:6869658 - 7138691 on Build GRCh38 | Gain |
|
| nsv510752 | Chr.19:6851075 - 6916782 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map