Assay Details
Target Gene Details
Entrez Gene ID: | 55709 |
Gene Name: | kelch repeat and BTB domain containing 4 |
Gene Aliases: |
BKLHD4, HSPC252 |
Location: |
Chr.11:47572197-47579015 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KBTBD4 | NM_001318716.1 | 2 | 424 | NP_001305645.1 |
| NM_001318717.1 | 2 | 345 | NP_001305646.1 | |
| NM_001318718.1 | 2 | 325 | NP_001305647.1 | |
| NM_001318719.1 | 2 | 303 | NP_001305648.1 | |
| NM_001318720.1 | 2 | 297 | NP_001305649.1 | |
| NM_001318721.1 | 2 | 272 | NP_001305650.1 | |
| NM_001318722.1 | 2 | 293 | NP_001305651.1 | |
| NM_001318723.1 | 2 | 377 | NP_001305652.1 | |
| NM_001318724.1 | 2 | 377 | NP_001305653.1 | |
| NM_001318725.1 | 2 | 280 | NP_001305654.1 | |
| NM_016506.6 | 2 | 381 | NP_057590.3 | |
| NM_018095.5 | 2 | 276 | NP_060565.4 | |
| XM_017018009.1 | 2 | 260 | XP_016873498.1 | |
| AF151086.1 | 2 | 238 | AAF36172.1 | |
| AK001312.1 | 2 | 230 | BAA91616.1 | |
| AK001749.1 | 2 | 317 | BAA91880.1 | |
| AK091607.1 | 1 | 954 | ||
| AK222550.1 | 2 | 227 | BAD96270.1 | |
| AK222985.1 | 2 | 237 | ||
| AK225062.1 | 2 | 227 | ||
| AL713735.1 | 1 | 206 | CAD28521.1 | |
| BC002736.2 | 2 | 318 | AAH02736.1 | |
| BI601113.1 | 2 | 284 | ||
| BI908078.1 | 2 | 283 | ||
| CR457270.1 | 1 | 164 | CAG33551.1 | |
| DA016320.1 | 2 | 276 | ||
| DA170281.1 | 2 | 332 | ||
| DA559313.1 | 2 | 335 | ||
| DA883418.1 | 2 | 314 | ||
| DB457928.1 | 2 | 248 | ||
| HY062454.1 | 2 | 259 | ||
| HY120885.1 | 2 | 258 | ||
| HY176105.1 | 2 | 300 | ||
| HY182004.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832142 | Chr.11:47496403 - 47659758 on Build GRCh38 | Loss |
|
| nsv825872 | Chr.11:47543873 - 47638778 on Build GRCh38 | Gain |
|
| nsv509405 | Chr.11:47571038 - 47663104 on Build GRCh38 | Insertion |
|
| nsv314 | Chr.11:47540012 - 47584704 on Build GRCh38 | Deletion |
|

Genomic Map