Assay Details
Target Gene Details
Entrez Gene ID: | 23256 |
Gene Name: | sec1 family domain containing 1 |
Gene Aliases: |
C14orf163, RA410, SLY1, SLY1P, STXBP1L2 |
Location: |
Chr.14:30622254-30735828 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv564169 | Chr.14:30349881 - 31072288 on Build GRCh38 | Gain |
|
| nsv564171 | Chr.14:30598884 - 30680424 on Build GRCh38 | Loss |
|
| nsv526536 | Chr.14:28322483 - 30669172 on Build GRCh38 | Gain |
|
| esv33189 | Chr.14:30331017 - 31442539 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map