Assay Details
Target Gene Details
Entrez Gene ID: | 23648 |
Gene Name: | single stranded DNA binding protein 3 |
Gene Aliases: |
CSDP, SSDP, SSDP1 |
Location: |
Chr.1:54225431-54413479 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SSBP3 | NM_001009955.3 | NP_001009955.1 | ||
| NM_018070.4 | NP_060540.2 | |||
| NM_145716.3 | NP_663768.1 | |||
| XM_006710545.3 | XP_006710608.1 | |||
| XM_017000897.1 | XP_016856386.1 | |||
| XM_017000898.1 | XP_016856387.1 | |||
| XM_017000899.1 | XP_016856388.1 | |||
| AF500116.1 | AAM22101.1 | |||
| AK001217.1 | BAA91561.1 | |||
| AK093975.1 | ||||
| AK289474.1 | ||||
| BC003605.1 | AAH03605.1 | |||
| BC004335.1 | AAH04335.2 | |||
| BC066365.1 | AAH66365.1 |
Target Gene Details
Entrez Gene ID: | 619518 |
Gene Name: | SSBP3 antisense RNA 1 |
Gene Aliases: |
C1orf191, MST128, MSTP128 |
Location: |
Chr.1:54236443-54239063 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SSBP3-AS1 | NR_103541.1 | 1 | 1947 | |
| AF176918.1 | 1 | 353 | AAQ13662.1 |
More Information
Set Membership: |
|

Genomic Map