Assay Details
Target Gene Details
Entrez Gene ID: | 121551 |
Gene Name: | BTB domain containing 11 |
Gene Aliases: |
ABTB2B |
Location: |
Chr.12:107317168-107659642 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BTBD11 | NM_001018072.1 | NP_001018082.1 | ||
| XM_011537910.2 | XP_011536212.1 | |||
| XM_017018807.1 | XP_016874296.1 | |||
| AI638039.1 | ||||
| AK122818.1 | ||||
| AK124835.1 | BAC85963.1 | |||
| BC146824.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1038935 | Chr.12:107314243 - 108077556 on Build GRCh38 | Gain |
|
| nsv872 | Chr.12:107295161 - 107328566 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map