Assay Details
Target Gene Details
Entrez Gene ID: | 126272 |
Gene Name: | EP300 interacting inhibitor of differentiation 2B |
Gene Aliases: |
EID-2B, EID-3 |
Location: |
Chr.19:39531533-39532867 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EID2B | NM_152361.2 | 1 | 72 | NP_689574.1 |
| AK096263.1 | BAC04743.1 | |||
| BC009725.1 | AAH09725.2 | |||
| DA382981.1 | 1 | 72 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv137930 | Chr.19:39528849 - 39537216 on Build GRCh38 | Deletion |
|
| nsv579509 | Chr.19:39514000 - 39642798 on Build GRCh38 | Gain |
|
| esv3644331 | Chr.19:39530314 - 39535353 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map