Assay Details
Target Gene Details
Entrez Gene ID: | 23138 |
Gene Name: | NEDD4 binding protein 3 |
Gene Aliases: |
LZTS4 |
Location: |
Chr.5:178113506-178127034 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| N4BP3 | NM_015111.1 | NP_055926.1 | ||
| XM_006714834.3 | XP_006714897.1 | |||
| XM_011534473.1 | XP_011532775.1 | |||
| XM_011534474.1 | XP_011532776.1 | |||
| AB002339.1 | ||||
| AK127224.1 | 1 | 391 | BAC86891.1 | |
| AK295663.1 | ||||
| BC053323.1 | AAH53323.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1019404 | Chr.5:178080079 - 178432088 on Build GRCh38 | Gain |
|
| nsv600392 | Chr.5:178078020 - 178145926 on Build GRCh38 | Loss |
|
| nsv950731 | Chr.5:178117400 - 178124399 on Build GRCh38 | Deletion |
|
| dgv5844n100 | Chr.5:177883324 - 178184647 on Build GRCh38 | Gain |
|
| nsv600391 | Chr.5:178061280 - 178122490 on Build GRCh38 | Loss |
|

Genomic Map