Assay Details
Target Gene Details
Entrez Gene ID: | 4792 |
Gene Name: | NFKB inhibitor alpha |
Gene Aliases: |
IKBA, MAD-3, NFKBI |
Location: |
Chr.14:35401510-35404754 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NFKBIA | NM_020529.2 | 6 | 1412 | NP_065390.1 |
| BC002601.2 | 6 | 1404 | AAH02601.1 | |
| BC004983.1 | 6 | 1395 | AAH04983.1 | |
| BU737609.1 | 1 | 168 | ||
| M69043.1 | 6 | 1396 | AAA16489.1 |
More Information
Set Membership: |
|

Genomic Map