Assay Details
Target Gene Details
Entrez Gene ID: | 25797 |
Gene Name: | glutaminyl-peptide cyclotransferase |
Gene Aliases: |
GCT, QC, sQC |
Location: |
Chr.2:37344610-37373322 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| QPCT | NM_012413.3 | 3 | 541 | NP_036545.1 |
| AK290605.1 | 3 | 523 | ||
| BC036721.1 | 3 | 420 | AAH36721.1 | |
| BC047756.2 | 3 | 518 | AAH47756.1 | |
| BI755122.1 | 3 | 576 | ||
| X67731.1 | 2 | 223 | CAA47961.1 | |
| X71125.1 | 3 | 430 | CAA50438.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2686 | Chr.2:37347524 - 37380543 on Build GRCh38 | Insertion |
|
| nsv2685 | Chr.2:37322953 - 37367780 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map