Assay Details
Target Gene Details
Entrez Gene ID: | 10642 |
Gene Name: | insulin like growth factor 2 mRNA binding protein 1 |
Gene Aliases: |
CRD-BP, CRDBP, IMP-1, IMP1, VICKZ1, ZBP1 |
Location: |
Chr.17:48997412-49056145 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IGF2BP1 | NM_001160423.1 | 1 | 89 | NP_001153895.1 |
| NM_006546.3 | 1 | 89 | NP_006537.3 | |
| XM_011524201.2 | XP_011522503.1 | |||
| XM_017024022.1 | XP_016879511.1 | |||
| AF198254.1 | 1 | 60 | AAF37203.1 | |
| AK313229.1 | 1 | 60 | ||
| BP238821.1 | 1 | 89 | ||
| DA758608.1 | 1 | 89 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953923 | Chr.17:48996139 - 49000138 on Build GRCh38 | Deletion |
|
| nsv833472 | Chr.17:48895840 - 49078381 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map