Assay Details
Target Gene Details
Entrez Gene ID: | 151613 |
Gene Name: | tetratricopeptide repeat domain 14 |
Gene Aliases: |
DRDL5813, PRO19630 |
Location: |
Chr.3:180602130-180618347 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC14 | NM_001042601.2 | NP_001036066.1 | ||
| NM_001288582.1 | NP_001275511.1 | |||
| NM_133462.3 | NP_597719.1 | |||
| XM_011512439.2 | XP_011510741.1 | |||
| XM_017005739.1 | XP_016861228.1 | |||
| XM_017005740.1 | XP_016861229.1 | |||
| AB075860.2 | BAB85566.2 | |||
| AK055984.1 | BAB71064.1 | |||
| AL832052.1 | CAH10404.1 | |||
| AL833086.2 | CAD89930.1 | |||
| AL833103.1 | ||||
| AY358762.1 | AAQ89122.1 | |||
| BC027990.1 | AAH27990.1 | |||
| BX648669.1 | 3 | 1848 |
More Information
Set Membership: |
|

Genomic Map