Assay Details
Target Gene Details
Entrez Gene ID: | 23185 |
Gene Name: | La ribonucleoprotein domain family member 4B |
Gene Aliases: |
KIAA0217, LARP5 |
Location: |
Chr.10:806914-931900 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LARP4B | NM_015155.2 | 18 | 5552 | NP_055970.1 |
| XM_017015987.1 | 17 | 7706 | XP_016871476.1 | |
| XM_017015988.1 | 17 | 7599 | XP_016871477.1 | |
| XM_017015989.1 | 17 | 7644 | XP_016871478.1 | |
| XM_017015992.1 | 13 | 4920 | XP_016871481.1 | |
| XM_017015993.1 | 16 | 5306 | XP_016871482.1 | |
| XM_017015994.1 | 17 | 5238 | XP_016871483.1 | |
| BC003381.1 | 1 | 209 | AAH03381.1 | |
| BC152443.1 | 17 | 5276 | ||
| D86971.2 | 17 | 5276 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv549753 | Chr.10:412537 - 848898 on Build GRCh38 | Gain |
|
| esv2660097 | Chr.10:526001 - 1246069 on Build GRCh38 | Deletion |
|
| nsv5498 | Chr.10:803197 - 814522 on Build GRCh38 | Deletion |
|
| nsv1048707 | Chr.10:788499 - 869594 on Build GRCh38 | Gain |
|
| nsv466698 | Chr.10:790175 - 842133 on Build GRCh38 | Loss |
|
| nsv549765 | Chr.10:688289 - 1083587 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map