Assay Details
Target Gene Details
Entrez Gene ID: | 57599 |
Gene Name: | WD repeat domain 48 |
Gene Aliases: |
P80, SPG60, UAF1 |
Location: |
Chr.3:39051986-39096671 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WDR48 | NM_001303402.1 | NP_001290331.1 | ||
| NM_001303403.1 | NP_001290332.1 | |||
| NM_020839.3 | NP_065890.1 | |||
| XM_005265346.3 | XP_005265403.1 | |||
| XM_011533971.1 | XP_011532273.1 | |||
| XM_017006960.1 | XP_016862449.1 | |||
| AB040882.2 | ||||
| AF468833.1 | AAL78650.1 | |||
| AK025513.1 | ||||
| AK297349.1 | ||||
| AK298810.1 | ||||
| AK302307.1 | ||||
| AL832926.1 | CAH56300.1 | |||
| BC026353.1 | AAH26353.1 | |||
| BC037168.2 | AAH37168.1 | |||
| DA215447.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1007100 | Chr.3:39051835 - 39094306 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map