Assay Details
Target Gene Details
Entrez Gene ID: | 121549 |
Gene Name: | achaete-scute family bHLH transcription factor 4 |
Gene Aliases: |
HASH4, bHLHa44 |
Location: |
Chr.12:107774385-107776644 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASCL4 | NM_203436.2 | 1 | 102 | NP_982260.2 |
| AY238895.1 | 1 | 102 | AAP69222.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1038935 | Chr.12:107314243 - 108077556 on Build GRCh38 | Gain |
|
| nsv560091 | Chr.12:107448429 - 108068129 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map