Assay Details
Target Gene Details
Entrez Gene ID: | 150921 |
Gene Name: | transcription factor 23 |
Gene Aliases: |
OUT, TCF-23, bHLHa24 |
Location: |
Chr.2:27148895-27153577 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TCF23 | NM_175769.2 | 2 | 441 | NP_786951.1 |
| XM_005264159.4 | 2 | 623 | XP_005264216.1 | |
| BC137191.1 | 2 | 441 | ||
| BC137192.1 | 2 | 441 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv6744n54 | Chr.2:26973431 - 27151141 on Build GRCh38 | Loss |
|
| nsv482050 | Chr.2:27149134 - 27152865 on Build GRCh38 | Gain |
|
| nsv817673 | Chr.2:27037601 - 27163931 on Build GRCh38 | Gain |
|
| nsv833603 | Chr.2:27059106 - 27263631 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map