Assay Details
Target Gene Details
Entrez Gene ID: | 5333 |
Gene Name: | phospholipase C delta 1 |
Gene Aliases: |
NDNC3, PLC-III |
Location: |
Chr.3:38007496-38029663 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLCD1 | NM_001130964.1 | 15 | 2655 | NP_001124436.1 |
| NM_006225.3 | 15 | 2362 | NP_006216.2 | |
| NR_024071.1 | 14 | 2732 | ||
| XM_017006622.1 | 15 | 2625 | XP_016862111.1 | |
| AB209862.1 | 15 | 2299 | BAD93099.1 | |
| AK090774.1 | 15 | 2444 | ||
| AK098690.1 | 12 | 1645 | ||
| AK127813.1 | 15 | 2272 | ||
| AK292324.1 | 15 | 2362 | ||
| BC050382.1 | 15 | 2326 | AAH50382.2 | |
| BX647927.1 | 14 | 2466 | CAI46087.1 | |
| U09117.1 | 15 | 2332 | AAA73567.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759140 | Chr.3:37789992 - 38062733 on Build GRCh38 | Loss |
|
| nsv590101 | Chr.3:38000691 - 38065526 on Build GRCh38 | Loss |
|
| nsv590094 | Chr.3:37941266 - 38021881 on Build GRCh38 | Loss |
|
| nsv522354 | Chr.3:37985093 - 38021881 on Build GRCh38 | Loss |
|
| nsv428416 | Chr.3:37864474 - 38062733 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map