Assay Details
Target Gene Details
Entrez Gene ID: | 167681 |
Gene Name: | protease, serine 35 |
Gene Aliases: |
C6orf158, dJ223E3.1 |
Location: |
Chr.6:83512475-83525704 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRSS35 | NM_001170423.1 | 3 | 1562 | NP_001163894.1 |
| NM_153362.2 | 2 | 1457 | NP_699193.2 | |
| AK291928.1 | 2 | 1342 | ||
| AY358661.1 | 3 | 1491 | AAQ89024.1 | |
| BC037170.1 | 2 | 1380 | AAH37170.1 |
More Information
Set Membership: |
|

Genomic Map