Assay Details
Target Gene Details
Entrez Gene ID: | 116179 |
Gene Name: | transglutaminase 7 |
Gene Aliases: |
TGMZ |
Location: |
Chr.15:43276277-43302255 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TGM7 | NM_052955.2 | 4 | 643 | NP_443187.1 |
| XM_017021903.1 | 4 | 641 | XP_016877392.1 | |
| AF363393.1 | 4 | 643 | AAK97573.1 |
More Information
Set Membership: |
|

Genomic Map