Assay Details
Target Gene Details
Entrez Gene ID: | 10092 |
Gene Name: | actin related protein 2/3 complex subunit 5 |
Gene Aliases: |
ARC16, dJ127C7.3, p16-Arc |
Location: |
Chr.1:183626193-183635941 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARPC5 | NM_001270439.1 | 4 | 839 | NP_001257368.1 |
| NM_005717.3 | 4 | 830 | NP_005708.1 | |
| AF006088.1 | 4 | 572 | AAB64193.1 | |
| AK095424.1 | 3 | 1210 | ||
| AK129598.1 | 4 | 615 | ||
| BC057237.1 | 4 | 659 | AAH57237.1 | |
| BI758106.1 | 4 | 740 |
More Information
Set Membership: |
|

Genomic Map