Assay Details
Target Gene Details
Entrez Gene ID: | 56181 |
Gene Name: | mitochondrial fission regulator 1 like |
Gene Aliases: |
FAM54B, HYST1888, MST116, MSTP116 |
Location: |
Chr.1:25819906-25832942 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTFR1L | NM_001099625.1 | NP_001093095.1 | ||
| NM_001099626.1 | NP_001093096.1 | |||
| NM_001099627.1 | NP_001093097.1 | |||
| NM_019557.5 | NP_062457.3 | |||
| AB075880.1 | BAD38662.1 | |||
| AF173891.1 | AAQ13638.1 | |||
| AK056721.1 | ||||
| AK290744.1 | ||||
| AK299221.1 | ||||
| AK314297.1 | ||||
| AL512766.1 | CAC21683.1 | |||
| AY007100.1 | ||||
| BC017175.2 | AAH17175.1 | |||
| BI545005.1 | ||||
| BI769886.1 | ||||
| BX647921.1 | CAH56194.1 | |||
| DB170641.1 | ||||
| KU178723.1 | ||||
| KU178724.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv7490 | Chr.1:25797857 - 25843537 on Build GRCh38 | Deletion |
|
| nsv545883 | Chr.1:25778482 - 25891593 on Build GRCh38 | Loss |
|
| nsv834491 | Chr.1:25726411 - 25914117 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map