Assay Details
Target Gene Details
Entrez Gene ID: | 284131 |
Gene Name: | endonuclease V |
Gene Aliases: |
- |
Location: |
Chr.17:80415117-80438086 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv24254 | Chr.17:80396528 - 80656857 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map