Assay Details
Target Gene Details
Entrez Gene ID: | 204801 |
Gene Name: | NLR family pyrin domain containing 11 |
Gene Aliases: |
CLR19.6, NALP11, NOD17, PAN10, PYPAF6, PYPAF7 |
Location: |
Chr.19:55785397-55836762 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NLRP11 | NM_001297743.1 | 9 | 2853 | NP_001284672.1 |
| NM_145007.3 | 12 | 3703 | NP_659444.2 | |
| AK090621.1 | 7 | 1741 | BAC03490.1 | |
| AK129784.1 | 7 | 1768 | ||
| AY095145.1 | 10 | 3091 | AAM14632.1 | |
| AY154466.1 | 9 | 2992 | AAO18162.1 | |
| AY168967.1 | 10 | 3080 | AAO12532.1 | |
| AY168968.1 | 9 | 2752 | AAO12533.1 | |
| AY226382.1 | 9 | 2992 | AAO59428.1 | |
| BC034730.1 | 10 | 3080 | AAH34730.1 | |
| BU622142.1 | 1 | 357 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1066324 | Chr.19:55754298 - 55789134 on Build GRCh38 | Gain+Loss |
|
| nsv1060544 | Chr.19:55751646 - 55797784 on Build GRCh38 | Gain |
|
| nsv833882 | Chr.19:55647267 - 55815198 on Build GRCh38 | Loss |
|
| esv2758773 | Chr.19:55724171 - 55814940 on Build GRCh38 | Gain |
|
| nsv580346 | Chr.19:55735154 - 55857948 on Build GRCh38 | Gain |
|
| nsv509753 | Chr.19:55738951 - 55799012 on Build GRCh38 | Insertion |
|
| nsv1060269 | Chr.19:55708879 - 55810275 on Build GRCh38 | Gain |
|
| nsv2556 | Chr.19:55745802 - 55799840 on Build GRCh38 | Insertion |
|
| nsv1057383 | Chr.19:55699665 - 55944662 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map