Assay Details
Target Gene Details
Entrez Gene ID: | 7942 |
Gene Name: | transcription factor EB |
Gene Aliases: |
ALPHATFEB, BHLHE35, TCFEB |
Location: |
Chr.6:41683978-41736259 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TFEB | NM_001167827.2 | 9 | 1540 | NP_001161299.2 |
| NM_001271943.1 | 8 | 1349 | NP_001258872.1 | |
| NM_001271944.1 | 9 | 1604 | NP_001258873.1 | |
| NM_001271945.1 | 9 | 1515 | NP_001258874.1 | |
| NM_007162.2 | 10 | 1706 | NP_009093.1 | |
| XM_005249411.1 | 9 | 1639 | XP_005249468.1 | |
| XM_005249412.1 | 9 | 1531 | XP_005249469.1 | |
| XM_006715212.3 | 9 | 1758 | XP_006715275.1 | |
| XM_006715213.2 | 9 | 1582 | XP_006715276.1 | |
| XM_011514915.1 | 9 | 1761 | XP_011513217.1 | |
| XM_011514916.2 | 10 | 1551 | XP_011513218.1 | |
| AJ608786.1 | 9 | 1540 | CAE77681.1 | |
| AK298390.1 | 9 | 1525 | ||
| BC006225.2 | 7 | 978 | AAH06225.2 | |
| BC032448.1 | 10 | 1706 | AAH32448.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759422 | Chr.6:41621051 - 41731715 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map