Assay Details
Target Gene Details
Entrez Gene ID: | 117286 |
Gene Name: | calcium and integrin binding family member 3 |
Gene Aliases: |
KIP3 |
Location: |
Chr.19:16161368-16173525 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CIB3 | NM_001300922.1 | NP_001287851.1 | ||
| NM_054113.3 | NP_473454.1 | |||
| AB050868.1 | BAB71789.1 | |||
| AW295492.1 | ||||
| BC069320.1 | AAH69320.2 | |||
| BC069428.1 | AAH69428.1 | |||
| BC069524.1 | AAH69524.1 | |||
| BC112200.1 | AAI12201.1 | |||
| BC113591.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3643791 | Chr.19:16149438 - 16170044 on Build GRCh38 | Loss |
|
| nsv833767 | Chr.19:16121391 - 16347760 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map