Assay Details
Target Gene Details
Entrez Gene ID: | 29994 |
Gene Name: | bromodomain adjacent to zinc finger domain 2B |
Gene Aliases: |
WALp4 |
Location: |
Chr.2:159315312-159712439 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BAZ2B | NM_001289975.1 | 1 | 206 | NP_001276904.1 |
| NM_013450.3 | 1 | 206 | NP_038478.2 | |
| XM_006712461.3 | 4 | 623 | XP_006712524.2 | |
| XM_011511038.2 | 1 | 153 | XP_011509340.1 | |
| XM_011511047.1 | 1 | 144 | XP_011509349.1 | |
| XM_017003918.1 | 4 | 613 | XP_016859407.1 | |
| XM_017003919.1 | 4 | 607 | XP_016859408.1 | |
| XM_017003920.1 | 5 | 716 | XP_016859409.1 | |
| XM_017003921.1 | 1 | 153 | XP_016859410.1 | |
| XM_017003925.1 | 1 | 153 | XP_016859414.1 | |
| XM_017003926.1 | 1 | 110 | XP_016859415.1 | |
| XM_017003927.1 | 1 | 144 | XP_016859416.1 | |
| XM_017003928.1 | 1 | 153 | XP_016859417.1 | |
| XM_017003931.1 | 1 | 153 | XP_016859420.1 | |
| XM_017003934.1 | 1 | 110 | XP_016859423.1 | |
| XM_017003935.1 | 1 | 144 | XP_016859424.1 | |
| XM_017003936.1 | 1 | 110 | XP_016859425.1 | |
| AB040909.2 | 1 | 174 | ||
| AL834381.1 | 1 | 101 | CAD39044.2 | |
| BX641037.1 | 4 | 598 | CAE46023.1 | |
| BX647250.1 | 1 | 55 |
Target Gene Details
Entrez Gene ID: | 643072 |
Gene Name: | uncharacterized LOC643072 |
Gene Aliases: |
- |
Location: |
Chr.2:159615294-159618042 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC643072 | NR_110587.1 | 2 | 810 | |
| NR_110588.1 | 2 | 807 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1010774 | Chr.2:159012247 - 160175703 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map