Assay Details
Target Gene Details
Entrez Gene ID: | 196883 |
Gene Name: | adenylate cyclase 4 |
Gene Aliases: |
AC4 |
Location: |
Chr.14:24318349-24335071 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADCY4 | NM_001198568.1 | NP_001185497.1 | ||
| NM_001198592.1 | NP_001185521.1 | |||
| NM_139247.3 | NP_640340.2 | |||
| AF497516.1 | AAM94373.1 | |||
| AK122714.1 | ||||
| AK126468.1 | BAC86560.1 | |||
| BC117473.1 | ||||
| BC117475.1 | ||||
| BX248285.1 | CAD62613.1 | |||
| BX537489.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv564060 | Chr.14:24307195 - 24340589 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map