Assay Details
Target Gene Details
Entrez Gene ID: | 80232 |
Gene Name: | WD repeat domain 26 |
Gene Aliases: |
CDW2, GID7, MIP2 |
Location: |
Chr.1:224385143-224434299 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WDR26 | NM_001115113.2 | 14 | 4250 | NP_001108585.2 |
| NM_025160.6 | 14 | 4298 | NP_079436.4 | |
| XM_005273273.4 | 13 | 4013 | XP_005273330.1 | |
| XM_006711817.3 | 14 | 4196 | XP_006711880.1 | |
| XM_011544281.2 | 14 | 4039 | XP_011542583.1 | |
| XM_011544282.2 | 11 | 3361 | XP_011542584.1 | |
| XM_017002396.1 | 14 | 3887 | XP_016857885.1 | |
| AB209887.1 | 14 | 4298 | BAD93124.1 | |
| AK023023.1 | 1 | 216 | ||
| AK027236.1 | 1 | 1503 | ||
| AY221751.1 | 14 | 4088 | AAO67709.1 | |
| BC063817.1 | 10 | 3360 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv8868 | Chr.1:224195321 - 224564829 on Build GRCh38 | Loss |
|
| nsv469889 | Chr.1:224252652 - 224446672 on Build GRCh38 | Gain |
|

Genomic Map