Assay Details
Target Gene Details
Entrez Gene ID: | 1540 |
Gene Name: | CYLD lysine 63 deubiquitinase |
Gene Aliases: |
BRSS, CDMT, CYLD1, CYLDI, EAC, MFT, MFT1, SBS, TEM, USPL2 |
Location: |
Chr.16:50742026-50801935 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYLD | NM_001042355.1 | 4 | 990 | NP_001035814.1 |
| NM_001042412.1 | 4 | 1014 | NP_001035877.1 | |
| NM_015247.2 | 5 | 1127 | NP_056062.1 | |
| XM_005255812.2 | 5 | 1087 | XP_005255869.1 | |
| XM_006721149.1 | 5 | 1151 | XP_006721212.1 | |
| XM_011522907.2 | 4 | 913 | XP_011521209.1 | |
| XM_011522908.1 | 4 | 1005 | XP_011521210.1 | |
| XM_011522911.2 | 4 | 1005 | XP_011521213.1 | |
| XM_017022977.1 | 4 | 1285 | XP_016878466.1 | |
| XM_017022978.1 | 4 | 1081 | XP_016878467.1 | |
| XM_017022979.1 | 4 | 1041 | XP_016878468.1 | |
| XM_017022980.1 | 4 | 1074 | XP_016878469.1 | |
| XM_017022981.1 | 4 | 972 | XP_016878470.1 | |
| AF161542.1 | 3 | 962 | AAF29029.1 | |
| AJ250014.1 | 5 | 1103 | CAB93533.1 | |
| AK024212.1 | 2 | 448 | ||
| AK024348.1 | 2 | 497 | ||
| AK056226.1 | 4 | 945 | ||
| AK225202.1 | 5 | 1107 | ||
| AK292975.1 | 4 | 972 | ||
| AK310832.1 | 4 | 962 | ||
| BC012342.1 | 4 | 988 | AAH12342.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952674 | Chr.16:50743190 - 50793689 on Build GRCh38 | Duplication |
|
| esv3638608 | Chr.16:50654254 - 50854693 on Build GRCh38 | Gain |
|
| dgv2977n100 | Chr.16:50744296 - 50798809 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map