Assay Details
Target Gene Details
Entrez Gene ID: | 84159 |
Gene Name: | AT-rich interaction domain 5B |
Gene Aliases: |
DESRT, MRF-2, MRF2 |
Location: |
Chr.10:61901254-62096948 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARID5B | NM_032199.2 | 2 | 572 | NP_115575.1 |
| XM_011540262.1 | 2 | 572 | XP_011538564.1 | |
| AK296921.1 | 2 | 138 | ||
| BC107800.1 | 2 | 131 | AAI07801.1 | |
| BG615173.1 | 2 | 143 | ||
| DB014195.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv8681 | Chr.10:61896798 - 62671529 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map